MAN424072894 Manchester Organics

2,4,6-tris(1,1,2,2,3,3,3-heptafluoropropyl)-1,3,5-triazine

  • MDL number MFCD00042438
  • Cas Number : 915-76-4
  • Molecular Weight : 585.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/F48881
Availability details for
Quantity
  • Availability
  • Add to Cart