MAN424073052 Manchester Organics

Methyl 5-chloro-6-methyl-pyrazine-2-carboxylate

  • MDL number MFCD18633146
  • Cas Number : 77168-85-5
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/M48956
Availability details for
Quantity
  • Availability
  • Add to Cart