MAN428715112 Manchester Organics

tert-Butyl (1S,5R)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD17016708
  • Molecular Weight : 212.28869

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J35958

Safety Information

MSDS https://www.manchesterorganics.com/J35958
Availability details for
Quantity
  • Availability
  • Add to Cart