MAN428715190 Manchester Organics

Morpholin-3-ylmethanol Hydrochloride

  • MDL number MFCD12547467
  • Molecular Weight : 153.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S23506

Safety Information

MSDS https://www.manchesterorganics.com/S23506
Availability details for
Quantity
  • Availability
  • Add to Cart