MAN428715191 Manchester Organics

(2S)-2-Acetamido-5-ureido-pentanoic acid

  • MDL number MFCD12407169
  • Molecular Weight : 217.222

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B49015
Availability details for
Quantity
  • Availability
  • Add to Cart