MAN428715291 Manchester Organics

Di(imidazol-1-yl)methanimine

  • MDL number MFCD10699388
  • Cas Number : 104619-51-4
  • Molecular Weight : 161.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/S49029
Availability details for
Quantity
  • Availability
  • Add to Cart