MAN428715520 Manchester Organics

Ethyl 8-bromooctanoate

  • MDL number MFCD00045057
  • Cas Number : 29823-21-0
  • Molecular Weight : 251.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 8-bromooctanoate
Inchi Code 1S/C10H19BrO2/c1-2-13-10(12)8-6-4-3-5-7-9-11/h2-9H2,1H3
Inchi Key UBTQVPMVWAEGAC-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/S49213
Availability details for
Quantity
  • Availability
  • Add to Cart