MAN428715534 Manchester Organics

Methyl 4-bromo-2-fluorobenzoate

  • MDL number MFCD06203731
  • Molecular Weight : 233.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/G49227
Availability details for
Quantity
  • Availability
  • Add to Cart