MAN428715650 Manchester Organics

1-Hydroxy-1,2-benziodoxol-3(1H)-one

  • MDL number MFCD01658875
  • Molecular Weight : 264.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T49306
Availability details for
Quantity
  • Availability
  • Add to Cart