MAN428715653 Manchester Organics

2-(2,6-Dibromophenyl)acetic acid

  • MDL number MFCD09832981
  • Cas Number : 901310-02-9
  • Molecular Weight : 293.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W49309
Availability details for
Quantity
  • Availability
  • Add to Cart