MAN428715709 Manchester Organics

1-(4-Chloro-2-pyridyl)ethanone

  • MDL number MFCD10697662
  • Molecular Weight : 155.58164

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K49322

Safety Information

MSDS https://www.manchesterorganics.com/K49322
Availability details for
Quantity
  • Availability
  • Add to Cart