MAN428716008 Manchester Organics

(4-Nitrophenyl) dihydrogen phosphate

  • MDL number MFCD00067095
  • Molecular Weight : 219.0887

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R49396

Safety Information

MSDS https://www.manchesterorganics.com/R49396
Availability details for
Quantity
  • Availability
  • Add to Cart