MAN428716136 Manchester Organics

2-(3-Fluoro-2-methyl-phenyl)acetic acid

  • MDL number MFCD03788545
  • Cas Number : 500912-16-3
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/D49316
Availability details for
Quantity
  • Availability
  • Add to Cart