MAN428716211 Manchester Organics

4-(Carboxymethyl)benzoic acid

  • MDL number MFCD01074834
  • Molecular Weight : 180.157

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S49581
Availability details for
Quantity
  • Availability
  • Add to Cart