MAN428716579 Manchester Organics

2-(2-Bromophenyl)-2,2-difluoro-ethanol

  • MDL number MFCD28038787
  • Molecular Weight : 237.041

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 0.94
Availability details for
Quantity
  • Availability
  • Add to Cart