MAN428716590 Manchester Organics

4-Bromo-3,5-difluoro-benzaldehyde

  • MDL number MFCD16659085
  • Molecular Weight : 220.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X49724
Availability details for
Quantity
  • Availability
  • Add to Cart