MAN432328718 Manchester Organics

2-Bromo-5-(trifluoromethyl)pyrimidine

  • MDL number MFCD14702304
  • Molecular Weight : 226.982

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E99666
Availability details for
Quantity
  • Availability
  • Add to Cart