MAN432328808 Manchester Organics

tert-Butyl (1S,5R)-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

  • MDL number MFCD17016736
  • Molecular Weight : 198.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z41193
Availability details for
Quantity
  • Availability
  • Add to Cart