MAN432329275 Manchester Organics

tert-Butyl N-[(3R,4R)-3-fluoro-4-piperidyl]carbamate

  • MDL number MFCD18632652
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/P49855
Availability details for
Quantity
  • Availability
  • Add to Cart