MAN432329350 Manchester Organics

2-Azaspiro[3.3]heptane-6-carboxylic acid trifluoroacetate

  • MDL number MFCD19982773
  • Cas Number : 1172252-57-1
  • Molecular Weight : 255.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T31918
Availability details for
Quantity
  • Availability
  • Add to Cart