MAN432329930 Manchester Organics

Cinnamylamine Hydrochloride

  • MDL number MFCD00184831
  • Cas Number : 5586-89-0
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/T98622
Availability details for
Quantity
  • Availability
  • Add to Cart