MAN432329937 Manchester Organics

O1-tert-Butyl O4-methyl 4-(aminomethyl)piperidine-1,4-dicarboxylate

  • MDL number MFCD07784122
  • Cas Number : 362703-35-3
  • Molecular Weight : 272.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-tert-butyl 4-methyl 4-(aminomethyl)-1,4-piperidinedicarboxylate
Inchi Code 1S/C13H24N2O4/c1-12(2,3)19-11(17)15-7-5-13(9-14,6-8-15)10(16)18-4/h5-9,14H2,1-4H3
Inchi Key HZHZQEZZXBLFHO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K49966
Availability details for
Quantity
  • Availability
  • Add to Cart