MAN432330104 Manchester Organics

4-Bromo-2-(4-bromo-2-pyridyl)pyridine

  • MDL number MFCD00667763
  • Cas Number : 18511-71-2
  • Molecular Weight : 313.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/J49988
Availability details for
Quantity
  • Availability
  • Add to Cart