MAN444778532 Manchester Organics

3-Fluoro-2-iodo-benzonitrile

  • MDL number MFCD07782075
  • Molecular Weight : 247.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P50039
Availability details for
Quantity
  • Availability
  • Add to Cart