MAN444778595 Manchester Organics

Fluoromethyl 4-methylbenzenesulfonate

  • MDL number MFCD20482732
  • Molecular Weight : 204.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V13001

Safety Information

MSDS https://www.manchesterorganics.com/V13001
Availability details for
Quantity
  • Availability
  • Add to Cart