MAN444778697 Manchester Organics

2,6-Dichloropyridin-3-ol

  • MDL number MFCD11044319
  • Molecular Weight : 163.989

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P50085
Availability details for
Quantity
  • Availability
  • Add to Cart