MAN444778701 Manchester Organics

tert-Butyl (2S)-2-propylpiperazine-1-carboxylate hydrochloride

  • MDL number MFCD07772097
  • Molecular Weight : 264.792

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M22247
Availability details for
Quantity
  • Availability
  • Add to Cart