MAN444778712 Manchester Organics

(3S)-3-Aminopyrrolidin-2-one hydrochloride

  • MDL number MFCD08273939
  • Cas Number : 56440-28-9
  • Molecular Weight : 136.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/K39639
Availability details for
Quantity
  • Availability
  • Add to Cart