MAN444778727 Manchester Organics

4-Cyclopropylaniline hydrochloride

  • MDL number MFCD23703139
  • Cas Number : 1588440-94-1
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C34480
Availability details for
Quantity
  • Availability
  • Add to Cart