MAN444778960 Manchester Organics

3-Amino-2-methoxy-pyridine-4-carboxylic acid

  • MDL number MFCD18262266
  • Cas Number : 870997-81-2
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L50105
Availability details for
Quantity
  • Availability
  • Add to Cart