MAN444778977 Manchester Organics

tert-Butyl 7-oxo-2-azaspiro[3.4]octane-2-carboxylate

  • MDL number MFCD22421762
  • Cas Number : 1363382-39-1
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/A50118
Availability details for
Quantity
  • Availability
  • Add to Cart