MAN444778978 Manchester Organics

4-Bromo-1,2-bis(trifluoromethyl)benzene

  • MDL number MFCD06253758
  • Cas Number : 320-29-6
  • Molecular Weight : 293.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D11115
Availability details for
Quantity
  • Availability
  • Add to Cart