MAN444779067 Manchester Organics

tert-Butyl 2,8-Diazaspiro[3.5]nonane-8-carboxylate hemioxalate

  • MDL number MFCD27987991
  • Cas Number : 1523618-26-9
  • Molecular Weight : 542.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y27417
Availability details for
Quantity
  • Availability
  • Add to Cart