MAN444779159 Manchester Organics

Iodo-1,1,2,2-tetrafluoroethane

  • MDL number MFCD00069111
  • Molecular Weight : 227.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C27515
Availability details for
Quantity
  • Availability
  • Add to Cart