MAN444779325 Manchester Organics

tert-Butyl 4-amino-3-fluoro-piperidine-1-carboxylate

  • MDL number MFCD10566556
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A50233
Availability details for
Quantity
  • Availability
  • Add to Cart