MAN444779328 Manchester Organics

(2R)-2-[(1S)-1,2-Dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one

  • MDL number MFCD00064328
  • Molecular Weight : 176.124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/D50236
Availability details for
Quantity
  • Availability
  • Add to Cart