MAN444779506 Manchester Organics

Methyl 1-aminocyclopentanecarboxylate hydrochloride

  • MDL number MFCD03840364
  • Molecular Weight : 179.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K27522
Availability details for
Quantity
  • Availability
  • Add to Cart