MAN444779547 Manchester Organics

5-Chloro-4-methyl-2-nitroaniline

  • MDL number MFCD00034066
  • Cas Number : 7149-80-6
  • Molecular Weight : 186.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z50278
Availability details for
Quantity
  • Availability
  • Add to Cart