MAN444779601 Manchester Organics

(2S)-2-(Fluoromethyl)pyrrolidine hydrochloride

  • MDL number MFCD07779382
  • Molecular Weight : 139.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R50293
Availability details for
Quantity
  • Availability
  • Add to Cart