MAN458950419 Manchester Organics

Ethyl 2-cyano-2-(2-trifluoromethyl-4H-chromene-4-ylidene)acetate

  • MDL number MFCD07780000
  • Molecular Weight : 309.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Y97856

Safety Information

MSDS https://www.manchesterorganics.com/Y97856
Availability details for
Quantity
  • Availability
  • Add to Cart