MAN458950631 Manchester Organics

N-Methyl-N-[(3R,4R)-4-methyl-3-piperidyl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine x hydrochloride

  • MDL number MFCD14706832
  • Molecular Weight : 281.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R50362
Availability details for
Quantity
  • Availability
  • Add to Cart