MAN458951069 Manchester Organics

3-Nitro-2-(trifluoromethyl)-2H-chromene

  • MDL number MFCD08273462
  • Molecular Weight : 245.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B97155

Safety Information

MSDS https://www.manchesterorganics.com/B97155
Availability details for
Quantity
  • Availability
  • Add to Cart