MAN458951132 Manchester Organics

6-(4-Fluorophenyl)-2-thioureido-4-(trifluoromethyl)pyrimidine

  • MDL number MFCD16614739
  • Molecular Weight : 316.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z98165

Safety Information

MSDS https://www.manchesterorganics.com/Z98165
Availability details for
Quantity
  • Availability
  • Add to Cart