MAN458951135 Manchester Organics

5-Bromo-2-(5-bromo-2-pyridyl)pyridine

  • MDL number MFCD06657883
  • Cas Number : 15862-18-7
  • Molecular Weight : 313.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5,5'-dibromo-2,2'-bipyridine
Inchi Code 1S/C10H6Br2N2/c11-7-1-3-9(13-5-7)10-4-2-8(12)6-14-10/h1-6H
Inchi Key JNWPRPLNUUMYCM-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/X50667
Availability details for
Quantity
  • Availability
  • Add to Cart