MAN458951167 Manchester Organics

6-(3-Fluorophenyl)-2-thioureido-4-(trifluoromethyl)pyrimidine

  • MDL number MFCD16614740
  • Molecular Weight : 316.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A97726

Safety Information

MSDS https://www.manchesterorganics.com/A97726
Availability details for
Quantity
  • Availability
  • Add to Cart