MAN458951224 Manchester Organics

5-Bromo-2-(3-chloro-2-pyridyl)pyrazole-3-carboxylic acid

  • MDL number MFCD08689880
  • Molecular Weight : 302.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/K50748
Availability details for
Quantity
  • Availability
  • Add to Cart