MAN458951321 Manchester Organics

[(2R)-Morpholin-2-yl]methanol hydrochloride

  • MDL number MFCD11052541
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L38950
Availability details for
Quantity
  • Availability
  • Add to Cart