MAN458951353 Manchester Organics

N-Benzylglycine

  • MDL number MFCD00190772
  • Cas Number : 17136-36-6
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (benzylamino)acetic acid
Inchi Code 1S/C9H11NO2/c11-9(12)7-10-6-8-4-2-1-3-5-8/h1-5,10H,6-7H2,(H,11,12)
Inchi Key KGSVNOLLROCJQM-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/R27575
Availability details for
Quantity
  • Availability
  • Add to Cart