MAN458951376 Manchester Organics

3-Chloro-2,5,6-trifluoro-pyridine

  • MDL number MFCD03001160
  • Cas Number : 2879-42-7
  • Molecular Weight : 167.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-2,5,6-trifluoropyridine
Inchi Code 1S/C5HClF3N/c6-2-1-3(7)5(9)10-4(2)8/h1H
Inchi Key OROKHXKJSDPRET-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/U50848
Availability details for
Quantity
  • Availability
  • Add to Cart