MAN458951402 Manchester Organics

(2R)-4-(tert-Butoxycarbonylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)butanoic acid

  • MDL number MFCD00798647
  • Molecular Weight : 440.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin GB
Product Link https://www.manchesterorganics.com/B50878
Availability details for
Quantity
  • Availability
  • Add to Cart